missing translation for 'onlineSavingsMsg'
Learn More

Imidazole, Mikromol™

Product Code. 18001310
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18001310 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18001310 Supplier Mikromol Supplier No. MM0015.02
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Imidazole
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Imidazole
CAS 288-32-4
Analyte or Component Names Imidazole
Shipping Condition Room Temperature
Molecular Formula C3 H4 N2
Certifications/Compliance ISO 17025
Synonym Bifonazole Imp. C (EP), Clotrimazole Imp. D (EP), Enalapril Maleate Imp. I (EP), Enalapril Imp. I (EP), Ondansetron Hydrochloride Dihydrate Imp. E (EP), Ondansetron Imp. E (EP), Sildenafil Citrate Imp. E (EP), Sildenafil Imp. E (EP), Imidazole, 1H-Imidazole, Zoledronic Acid Monohydrate Imp. C (Pharmeuropa), CCI 625, Ph Eur Clotrimazole Impurity D, Ph Eur Ondansetron Impurity E, Ph Eur Zanamivir Impurity G, Imidazole (CCI 625), GR 152919X, GR 212339X, GR 228446X, USP Zanamivir Related Compounds Mixture, Bifonazole Impurity C, Clotrimazole Impurity D, Enalapril Maleate Impurity I, Ondansetron Hydrochloride Dihydrate Impurity E, Sildenafil Citrate Impurity E, Zoledronic Acid Impurity C, Enalapril Impurity I, Ondansetron Impurity E, Sildenafil Impurity E
InChI Formula InChI=1S/C3H4N2/c1-2-5-3-4-1/h1-3H,(H,4,5)
SMILES c1c[nH]cn1
IUPAC Name 1H-imidazole
Molecular Weight (g/mol) 68.08
Quantity 100 mg
Formula Weight 68.0374 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.