missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin 11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB100-56484
This item is not returnable.
View return policy
Description
Importin 11 Polyclonal specifically detects Importin 11 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot.
Specifications
| Importin 11 | |
| Polyclonal | |
| Unconjugated | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| imp11, importin 11, Ran binding protein 11, Ran-binding protein 11, RANBP11, RanBP11importin-11, SLRN | |
| Rabbit | |
| 112 kDa | |
| 0.1 mg | |
| Primary | |
| Bovine, Chimpanzee, Dog, Human, Mouse, Rat, Rhesus Monkey. Predicted cross-reactivity based on sequence identity: Xenopus (94%). | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.5-2 ug/ml | |
| Q9UI26 | |
| IPO11 | |
| A portion of amino acids 1-50 of human Importin-11 was used as the immunogen (NP_001128251). | |
| Affinity Purified | |
| RUO | |
| 51194 | |
| Human, Mouse, Rat, Bovine, Canine, Primate | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction