missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin 11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
199.00 € - 358.00 €
Specifications
| Antigen | Importin 11 |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5-2 ug/ml |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18005424
|
Novus Biologicals
NB100-56484SS |
0.025 mg |
199.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18756593
|
Novus Biologicals
NB100-56484 |
0.1 mg |
358.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Importin 11 Polyclonal specifically detects Importin 11 in Human, Mouse, Rat, Bovine, Canine, Primate samples. It is validated for Western Blot.Specifications
| Importin 11 | |
| Western Blot 0.5-2 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat, Bovine, Canine, Primate | |
| Q9UI26 | |
| 51194 | |
| A portion of amino acids 1-50 of human Importin-11 was used as the immunogen (NP_001128251). | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 112 kDa |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS containing 0.2% gelatin with 0.05% Sodium Azide | |
| imp11, importin 11, Ran binding protein 11, Ran-binding protein 11, RANBP11, RanBP11importin-11, SLRN | |
| IPO11 | |
| IgG | |
| Affinity Purified | |
| Bovine, Chimpanzee, Dog, Human, Mouse, Rat, Rhesus Monkey. Predicted cross-reactivity based on sequence identity: Xenopus (94%). |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title