Learn More
Importin-8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
Brand:  Novus Biologicals NBP2-24751SS
 View more versions of this product
Product Code. 18083244
  • 231.00 € / 25µg
Description
Importin-8 Polyclonal specifically detects Importin-8 in Human, Primate samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Importin-8
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
FLJ21954, FLJ26580, Imp8, importin 8, importin-8, RAN binding protein 8, Ran-binding protein 8, RanBP8, RANBP8imp8
Rabbit
Protein G purified
RUO
10526
Human, Primate
Purified
Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
1.0 mg/ml
Western Blot 1:200-1:2000, Simple Western 1:20, Immunocytochemistry/Immunofluorescence 1:20-1:2000, Immunoprecipitation reported in scientific literature (PMID 19131589)
O15397
IPO8
A portion of amino acids 850-900 of human Importin-8 was used as immunogen for the antibody.
0.025 mg
Primary
The amino acid sequence used as immunogen for the POU3F2 antibody is 100% homologous in human, horse, pig, mouse, cattle, rat and dog. Alternative initiation produces three different forms of this protein (approximately 47, 29 and 27 kD).
Store at -20C. Avoid freeze-thaw cycles.