missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin-8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
231.00 € - 509.00 €
Specifications
| Antigen | Importin-8 |
|---|---|
| Concentration | 1.0 mg/ml |
| Dilution | Western Blot 1:200-1:2000, Simple Western 1:20, Immunocytochemistry/Immunofluorescence 1:20-1:2000, Immunoprecipitation reported in scientific literature (PMID 19131589) |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18083244
|
Novus Biologicals
NBP2-24751SS |
0.025 mg |
231.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18707643
|
Novus Biologicals
NBP2-24751 |
0.1 mg |
509.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Importin-8 Polyclonal specifically detects Importin-8 in Human, Primate samples. It is validated for Western Blot, Simple Western, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
| Importin-8 | |
| Western Blot 1:200-1:2000, Simple Western 1:20, Immunocytochemistry/Immunofluorescence 1:20-1:2000, Immunoprecipitation reported in scientific literature (PMID 19131589) | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| FLJ21954, FLJ26580, Imp8, importin 8, importin-8, RAN binding protein 8, Ran-binding protein 8, RanBP8, RANBP8imp8 | |
| IPO8 | |
| IgG | |
| Protein G purified | |
| The amino acid sequence used as immunogen for the POU3F2 antibody is 100% homologous in human, horse, pig, mouse, cattle, rat and dog. Alternative initiation produces three different forms of this protein (approximately 47, 29 and 27 kD). |
| 1.0 mg/ml | |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Human, Primate | |
| O15397 | |
| 10526 | |
| A portion of amino acids 850-900 of human Importin-8 was used as immunogen for the antibody. | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title