missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Importin alpha 3/KPNA4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
189.00 € - 544.00 €
Specifications
| Antigen | Importin alpha 3/KPNA4 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 2.5-20 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18525463
|
Novus Biologicals
NBP1-76300 |
0.1 mg |
544.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18536074
|
Novus Biologicals
NBP1-76300-0.025mg |
0.025 mg |
189.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Importin alpha 3/KPNA4 Polyclonal antibody specifically detects Importin alpha 3/KPNA4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Importin alpha 3/KPNA4 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 2.5-20 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS | |
| importin alpha 3, Importin alpha Q1, importin subunit alpha-4, importin-alpha-Q1, IPOA3, karyopherin alpha 4 (importin alpha 3), MGC12217, MGC26703, Qip1, QIP1Karyopherin subunit alpha-4, SRP3 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the amino terminus of human KPNA4. The immunogen is located within amino acids 30 - 80 of KPNA4. Amino Acid Squence: RNVPQEDICEDS | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| EAW78632 | |
| 3840 | |
| IgG | |
| Peptide affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title