missing translation for 'onlineSavingsMsg'
Learn More

Indometacin 1.0 mg/ml in Acetonitrile, LoGiCal

Product Code. 30461877
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461877 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461877 Supplier LoGiCal Supplier No. LGCAMP0584.0011
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Indomethacin
CAS 53-86-1
Analyte or Component Names Indomethacin
Shipping Condition Room Temperature
Molecular Formula C19 H16 Cl N O4
Certifications/Compliance ISO 17034
Synonym 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid, 1-(p-Chlorobenzoyl)-5-methoxy-2-methyl-indole-3-acetic acid, 1-(4-Chlorobenzoyl)-2-methyl-5-methoxyindole-3-acetic acid, 1-(4-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic acid, 1-(p-Chlorobenzoyl)-2-methyl-5-methoxy-3-indolylacetic acid, 1-(p-Chlorobenzoyl)-2-methyl-5-methoxyindole-3-acetic acid, 1-(p-Chlorobenzoyl)-5-methoxy-2-methyl-3-indolylacetic acid, 1-(p-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic acid, Amuno, Argun, Artracin, Artrinovo, Artrivia, Bonidin, Bonidon Gel, Catlep, Chibro-Amuno, Chrono-Indicid, Chrono-Indocid 75, Confortid, Dolcidium, Dolcidium PL, Dolovin, Durametacin, Elmetacin, Endol, Flam, Globa, Idomethine, Imbrilon, Inacid, Indacin, Indo-Phlogont, Indo-Rectolmin, Indo-Tablinen, IndoRich, Indocid, Indocid (pharmaceutical), Indocin, Indocollyre, Indomecol, Indomed, Indomee, Indometacin, Indometacine, Indomethacin, Indomethacine, Indomethine, Indomod, Indonol, Indoptic, Indoptol, Indorektal, Indoxen, Indren, Inflazon, Infrocin, Innamit, Inteban, Inteban SP, Lausit, Metacen, Metartril, Methazine, Metindol, Mezolin, Mikametan, Mobilan, N-(p-Chlorobenzoyl)-2-methyl-5-methoxy-3-indolylacetic acid, NSC 77541, Reumacide, Rheumacin LA, Sadoreum, Tannex, Vital Vitacid, Vonum, [1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetic acid, alpha-[1-(p-Chlorobenzoyl)-2-methyl-5-methoxy-3-indolyl]acetic acid, Acemetacin Impurity B
InChI Formula InChI=1S/C19H16ClNO4/c1-11-15(10-18(22)23)16-9-14(25-2)7-8-17(16)21(11)19(24)12-3-5-13(20)6-4-12/h3-9H,10H2,1-2H3,(H,22,23)
Solution Type Single Solution
SMILES COc1ccc2c(c1)c(CC(=O)O)c(C)n2C(=O)c3ccc(Cl)cc3
IUPAC Name 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
Solvent or Matrix Acetonitrile
Industry Type Clinical Testing
Molecular Weight (g/mol) 357.79
Quantity 1 mL
Formula Weight 357.0768 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.