missing translation for 'onlineSavingsMsg'
Learn More

Indometacin, Mikromol™

Product Code. 18050480
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18050480 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18050480 Supplier Mikromol Supplier No. MM0584.00
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Indometacin
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Indomethacin
CAS 53-86-1
Analyte or Component Names Indomethacin
Shipping Condition Room Temperature
Molecular Formula C19 H16 Cl N O4
Certifications/Compliance ISO 17025
Synonym 1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indole-3-acetic acid, 1-(p-Chlorobenzoyl)-5-methoxy-2-methyl-indole-3-acetic acid, 1-(4-Chlorobenzoyl)-2-methyl-5-methoxyindole-3-acetic acid, 1-(4-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic acid, 1-(p-Chlorobenzoyl)-2-methyl-5-methoxy-3-indolylacetic acid, 1-(p-Chlorobenzoyl)-2-methyl-5-methoxyindole-3-acetic acid, 1-(p-Chlorobenzoyl)-5-methoxy-2-methyl-3-indolylacetic acid, 1-(p-Chlorobenzoyl)-5-methoxy-2-methylindole-3-acetic acid, Amuno, Argun, Artracin, Artrinovo, Artrivia, Bonidin, Bonidon Gel, Catlep, Chibro-Amuno, Chrono-Indicid, Chrono-Indocid 75, Confortid, Dolcidium, Dolcidium PL, Dolovin, Durametacin, Elmetacin, Endol, Flam, Globa, Idomethine, Imbrilon, Inacid, Indacin, Indo-Phlogont, Indo-Rectolmin, Indo-Tablinen, IndoRich, Indocid, Indocid (pharmaceutical), Indocin, Indocollyre, Indomecol, Indomed, Indomee, Indometacin, Indometacine, Indomethacin, Indomethacine, Indomethine, Indomod, Indonol, Indoptic, Indoptol, Indorektal, Indoxen, Indren, Inflazon, Infrocin, Innamit, Inteban, Inteban SP, Lausit, Metacen, Metartril, Methazine, Metindol, Mezolin, Mikametan, Mobilan, N-(p-Chlorobenzoyl)-2-methyl-5-methoxy-3-indolylacetic acid, NSC 77541, Reumacide, Rheumacin LA, Sadoreum, Tannex, Vital Vitacid, Vonum, [1-(4-Chlorobenzoyl)-5-methoxy-2-methyl-1H-indol-3-yl]acetic acid, alpha-[1-(p-Chlorobenzoyl)-2-methyl-5-methoxy-3-indolyl]acetic acid, Acemetacin Impurity B
InChI Formula InChI=1S/C19H16ClNO4/c1-11-15(10-18(22)23)16-9-14(25-2)7-8-17(16)21(11)19(24)12-3-5-13(20)6-4-12/h3-9H,10H2,1-2H3,(H,22,23)
SMILES COc1ccc2c(c1)c(CC(=O)O)c(C)n2C(=O)c3ccc(Cl)cc3
IUPAC Name 2-[1-(4-chlorobenzoyl)-5-methoxy-2-methylindol-3-yl]acetic acid
Molecular Weight (g/mol) 357.79
Quantity 500 mg
Formula Weight 357.0768 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.