Learn More
Influenza A Hemagglutinin H3 Antibody (InA227), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB100-73161AF350
 View more versions of this product
Product Code. 18667459
  • 553.00 € / 0.20mL
Description
Influenza A Hemagglutinin H3 Monoclonal antibody specifically detects Influenza A Hemagglutinin H3 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Influenza A Hemagglutinin H3
Monoclonal
Alexa Fluor 350
Mouse
Protein A purified
RUO
Virus
Purified
Western Blot
InA227
50mM Sodium Borate
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/Shangdong/9/93 (strain H3N2).
0.2 mL
Primary
Store at 4°C in the dark.
IgG1