missing translation for 'onlineSavingsMsg'
Learn More

Influenza A Hemagglutinin H3 Antibody (InA227), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody

553.00 €

Specifications

Antigen Influenza A Hemagglutinin H3
Clone InA227
Applications Western Blot
Classification Monoclonal
Conjugate Alexa Fluor 350
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18667459
View Documents
Novus Biologicals
NB100-73161AF350
0.2 mL
553.00 €
0.20mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Influenza A Hemagglutinin H3 Monoclonal antibody specifically detects Influenza A Hemagglutinin H3 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

Influenza A Hemagglutinin H3
Western Blot
Alexa Fluor 350
Mouse
Virus
Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with Influenza A/Shangdong/9/93 (strain H3N2).
Primary
Store at 4°C in the dark.
InA227
Monoclonal
Purified
RUO
50mM Sodium Borate
IgG1
Protein A purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.