missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inorganic Pyrophosphatase/PPA1 Mouse anti-Human, Clone: 4G4, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59471-50UL
This item is not returnable.
View return policy
Description
Inorganic Pyrophosphatase/PPA1 Monoclonal specifically detects Inorganic Pyrophosphatase/PPA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| Inorganic Pyrophosphatase/PPA1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1071, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| diphosphate phosphohydrolase, EC 3.6.1.1, inorganic diphosphatase, inorganic pyrophosphatase, inorganic pyrophosphatase 1, IOPPPMGC111556, PP1, PPase, PPcytosolic inorganic pyrophosphatase, pyrophosphatase (inorganic), pyrophosphatase (inorganic) 1, pyrophosphatase 1, Pyrophosphate phospho-hydrolase, SID6-8061 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 5464 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b λ |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 4G4 | |
| Unconjugated | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| PPA1 | |
| Recombinant human PPA1 (1-289aa) purified from E. coli | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction