missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inorganic Pyrophosphatase/PPA1 Mouse anti-Human, Clone: 4G4, Novus Biologicals™
Mouse Monoclonal Antibody
304.00 € - 486.00 €
Specifications
| Antigen | Inorganic Pyrophosphatase/PPA1 |
|---|---|
| Clone | 4G4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1071, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18284321
|
Novus Biologicals
NBP2-59471-50UL |
50 μL |
304.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18238604
|
Novus Biologicals
NBP2-59471-100UL |
100 μL |
486.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Inorganic Pyrophosphatase/PPA1 Monoclonal specifically detects Inorganic Pyrophosphatase/PPA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Inorganic Pyrophosphatase/PPA1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| diphosphate phosphohydrolase, EC 3.6.1.1, inorganic diphosphatase, inorganic pyrophosphatase, inorganic pyrophosphatase 1, IOPPPMGC111556, PP1, PPase, PPcytosolic inorganic pyrophosphatase, pyrophosphatase (inorganic), pyrophosphatase (inorganic) 1, pyrophosphatase 1, Pyrophosphate phospho-hydrolase, SID6-8061 | |
| PPA1 | |
| IgG2b λ | |
| Protein A purified |
| 4G4 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1071, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 5464 | |
| Recombinant human PPA1 (1-289aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title