missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Inositol Nicotinate, LoGiCal
Specifications
Specifications
| Chemical Name or Material | Inositol Nicotinate |
| CAS | 6556-11-2 |
| Analyte or Component Names | Inositol Nicotinate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C42 H30 N6 O12 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Inositol Nicotinate, myo-Inositol, hexa-3-pyridinecarboxylate, Inositol, hexanicotinate, myo- (7CI,8CI), Nicotinic acid, hexaester with myo-inositol (6CI), 3-Pyridinecarboxylic acid, 1,2,3,4,5,6-cyclohexanehexayl ester, Dilcit, Dilexpal, Esantene, Hamovannid, Hexanicit, Hexanicotinoyl inositol, Hexanicotol, Hexopal, Inositol hexaniacinate, Inositol hexanicotinate, Inositol niacinate, Inositol nicotinate, Linodil, Meso-inositol hexanicotinate, Mesonex, Mesotal, Palohex, meso-Inositol hexanicotinate, myo-Inositol hexanicotinate |
| InChI Formula | InChI=1S/C42H30N6O12/c49-37(25-7-1-13-43-19-25)55-31-32(56-38(50)26-8-2-14-44-20-26)34(58-40(52)28-10-4-16-46-22-28)36(60-42(54)30-12-6-18-48-24-30)35(59-41(53)29-11-5-17-47-23-29)33(31)57-39(51)27-9-3-15-45-21-27/h1-24,31-36H/t31-,32-,33-,34+,35-,36- |
| Solution Type | Neat |
| SMILES | O=C(O[C@@H]1[C@@H](OC(=O)c2cccnc2)[C@H](OC(=O)c3cccnc3)[C@@H](OC(=O)c4cccnc4)[C@@H](OC(=O)c5cccnc5)[C@H]1OC(=O)c6cccnc6)c7cccnc7 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?