missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (01), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-23494-100ul
This item is not returnable.
View return policy
Description
Insulin Monoclonal antibody specifically detects Insulin in Human samples. It is validated for ELISA
Specifications
| Insulin | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 1 | |
| ELISA 1:1000-1:2000, Sandwich ELISA Capture 1:250-1:2000 | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Insulin (Uniprot#: P01308; Phe25-Lys53 & Gly90-Asn110). | |
| 100 μg | |
| Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| 3630 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction