missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody (01), Novus Biologicals™
Mouse Monoclonal Antibody
339.00 €
Specifications
| Antigen | Insulin |
|---|---|
| Clone | 1 |
| Dilution | ELISA 1:1000-1:2000, Sandwich ELISA Capture 1:250-1:2000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18662325
|
Novus Biologicals
NBP3-23494-100ul |
100 μg |
339.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18640184
|
Novus Biologicals
NBP3-23494-20ul |
20 μg |
339.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Insulin Monoclonal antibody specifically detects Insulin in Human samples. It is validated for ELISASpecifications
| Insulin | |
| ELISA 1:1000-1:2000, Sandwich ELISA Capture 1:250-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Insulin (Uniprot#: P01308; Phe25-Lys53 & Gly90-Asn110). | |
| Protein A purified |
| 1 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Diabetes Research, Immune System Diseases, Immunology, Lipid and Metabolism, Stem Cell Markers | |
| 0.2 um filtered solution in PBS | |
| 3630 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title