missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-45662-0.025MG
This item is not returnable.
View return policy
Description
Insulin Polyclonal specifically detects Insulin in Human samples. It is validated for Western Blot.
Specifications
| Insulin | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2), 0.5% BSA, 30% glycerol with 0.01% Thimerosal | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 0.5 mg/mL | |
| Western Blot 0.5 - 4 ug/mL | |
| P01308 | |
| INS | |
| E. coli expressed recombinant human Insulin. | |
| 0.025 mg | |
| Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers | |
| 3630 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction