missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Insulin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
230.00 € - 570.00 €
Specifications
| Antigen | Insulin |
|---|---|
| Concentration | 0.5 mg/mL |
| Dilution | Western Blot 0.5 - 4 ug/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18221314
|
Novus Biologicals
NBP1-45662-0.025MG |
0.025 mg |
230.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18745963
|
Novus Biologicals
NBP1-45662 |
570.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
Insulin Polyclonal specifically detects Insulin in Human samples. It is validated for Western Blot.Specifications
| Insulin | |
| Western Blot 0.5 - 4 ug/mL | |
| Polyclonal | |
| Rabbit | |
| Diabetes Research, Immune System Diseases, Immunology, Stem Cell Markers | |
| PBS (pH 7.2), 0.5% BSA, 30% glycerol with 0.01% Thimerosal | |
| IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin | |
| INS | |
| IgG | |
| Affinity Purified |
| 0.5 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| P01308 | |
| 3630 | |
| E. coli expressed recombinant human Insulin. | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title