missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 1/CD49a Antibody (CL7207) - Azide and BSA Free, Novus Biologicals™

Product Code. 30287255 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30287255 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30287255 missing translation for 'mfr' Novus Biologicals missing translation for 'supplierNo' NBP344430

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Integrin alpha 1/CD49a Monoclonal antibody specifically detects Integrin alpha 1/CD49a in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin),Immunohistochemistry,KnockDown,Neutralization
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha 1/CD49a
Applications Western Blot, Immunohistochemistry (Paraffin), Immunohistochemistry, KnockDown, Neutralization
Classification Monoclonal
Clone CL7207
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry-Paraffin 1:1000 - 1:2500
Formulation Lyophilized in 5% Trehalose.
Gene Alias CD49 antigen-like family member A, CD49a, CD49a antigen, integrin, alpha 1, Laminin and collagen receptor, very late activation protein 1, VLA-1, VLA1integrin alpha-1
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of P56199, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 3672
Reconstitution Centrifuge the vial of lyophilized antibody at 12,000 x g for 20 seconds. Reconstitute by adding sterile, distilled water to achieve a final antibody concentration of 1mg/mL.
Target Species Human
Content And Storage Store at 4⪚C short term. Aliquot and store at -20⪚C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.