missing translation for 'onlineSavingsMsg'
Learn More

Integrin alpha 2b/CD41 Antibody (M148) - BSA Free, Novus Biologicals™

Product Code. 18718753 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18718753 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18718753 Supplier Novus Biologicals Supplier No. NB1002614

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Integrin alpha 2b/CD41 Monoclonal specifically detects Integrin alpha 2b/CD41 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunofluorescence, CyTOF-ready.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha 2b/CD41
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunofluorescence, CyTOF, CyTOF
Classification Monoclonal
Clone M148
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:10 - 1:1000, Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, Immunofluorescence 1:10 - 1:500, CyTOF-ready
Gene Alias CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK
Gene Symbols ITGA2B
Host Species Mouse
Immunogen Homogenized human medulloblastoma tissue.
Purification Method Protein A purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3674
Test Specificity M148 binds to some medulloblastoma and neuroblastoma cells and rhabdomyosarcoma and certain other solid tumours. No binding observed to marrow constituents other than platelets and megakaryocytes.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.