missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Integrin beta 8 Monoclonal antibody specifically detects Integrin beta 8 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Integrin beta 8 |
| Applications | Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Monoclonal |
| Clone | CL7290 |
| Conjugate | PE |
| Formulation | PBS |
| Gene Alias | integrin beta-8, integrin, beta 8 |
| Host Species | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from P26012, with the exact immunogen sequence remaining proprietary. |
| Purification Method | Protein A purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?