Learn More
Iodobenzene, 98%
Description
Iodobenzene is used to prepare biphenyl and stilbene. It reacts with magnesium to form the Grignard reagent, a phenylmagnesium iodide used in organic synthesis. It finds application as a solid chlorine source by reacting with chlorine to form a complex, iodobenzene dichloride. It serves as a substrate in Sonogashira reaction which is used to form a carbon-carbon bond between a terminal alkyne and an aryl halide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Iodobenzene is used to prepare biphenyl and stilbene. It reacts with magnesium to form the Grignard reagent, a phenylmagnesium iodide used in organic synthesis. It finds application as a solid chlorine source by reacting with chlorine to form a complex, iodobenzene dichloride. It serves as a substrate in Sonogashira reaction which is used to form a carbon-carbon bond between a terminal alkyne and an aryl halide.
Solubility
Miscible with ethanol, chloroform and ether. Slightly miscible with dimethyl sulfoxide. Immiscible with water.
Notes
Light sensitive. Incompatible with magnesium and strong oxidizing agents.
Spécification
Spécification
| Melting Point | °C to 31°C |
| Density | 1.826 |
| Boiling Point | 188°C to 189°C |
| Flash Point | 74°C (165°F) |
| Assay Percent Range | 98% |
| Refractive Index | 1.62 |
| Quantity | 50 g |
| Beilstein | 1446140 |
| Sensitivity | Light sensitive |
| Merck Index | 14,5029 |
| Afficher plus |
Sécurité et traitement
- Iodobenzene
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contain: Iodobenzene
RUO – Research Use Only
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.