missing translation for 'onlineSavingsMsg'
Learn More

Iodobenzene, 98%

Product Code. 15760799 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15760799 500 mL Glass bottle 500mL
10558850 100 mL Glass bottle 100mL
11914521 2.5 L Glass bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15760799 Supplier Thermo Scientific Acros Supplier No. 122335000
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 591-50-4 | C6H5I | 204.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 591-50-4
Molecular Formula C6H5I
Molecular Weight (g/mol) 204.01
MDL Number MFCD00001029
InChI Key SNHMUERNLJLMHN-UHFFFAOYSA-N
Synonym benzene, iodo, phenyl iodide, benzene iodide, iodophenyl, iodinebenzol, iodo-benzene, phenyliodide, iodo benzene, unii-9hk5l7ybbr, 1-iodobenzene
PubChem CID 11575
IUPAC Name iodobenzene
SMILES IC1=CC=CC=C1

Specifications

Melting Point -29.0°C
Color Yellow
Density 1.8200g/mL
Boiling Point 188.0°C
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5I
Refractive Index 1.6180 to 1.6200
Quantity 500 mL
Beilstein ,215
Fieser 01,505
Merck Index 15,5072
Specific Gravity 1.82
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether
Formula Weight 204.01
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Iodobenzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.