Learn More
IRF4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-76574
 View more versions of this product
Product Code. 18555893
  • 518.00 € / 0.10mg
Description
IRF4 Polyclonal antibody specifically detects IRF4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IRF4
Polyclonal
Unconjugated
PBS
interferon regulatory factor 4, IRF-4, LSIRFMUM1, Lymphocyte-specific interferon regulatory factor, Multiple myeloma oncogene 1, NF-EM5
Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human IRF4. The immunogen is located within the last 50 amino acids of IRF4. Amino Acid Squence: NPEDYHRSIRHSSIQE
0.1 mg
Cancer
3662
Store at 4°C.
IgG
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
1 mg/mL
Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 10-20 μg/mL
NP_002451
Rabbit
Peptide affinity purified
RUO
Primary
Human, Mouse, Rat
Purified