missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IRF4 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
178.00 € - 518.00 €
Specifications
| Antigen | IRF4 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 10-20 μg/mL |
| Applications | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18504074
|
Novus Biologicals
NBP1-76574-0.025mg |
0.025 mg |
178.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18555893
|
Novus Biologicals
NBP1-76574 |
0.1 mg |
518.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IRF4 Polyclonal antibody specifically detects IRF4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| IRF4 | |
| Western Blot 0.5 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/ Immunofluorescence 10-20 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| NP_002451 | |
| 3662 | |
| IgG | |
| Peptide affinity purified |
| 1 mg/mL | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS | |
| interferon regulatory factor 4, IRF-4, LSIRFMUM1, Lymphocyte-specific interferon regulatory factor, Multiple myeloma oncogene 1, NF-EM5 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human IRF4. The immunogen is located within the last 50 amino acids of IRF4. Amino Acid Squence: NPEDYHRSIRHSSIQE | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title