missing translation for 'onlineSavingsMsg'
Learn More

Isoleucine (L-Isoleucine), LoGiCal

Product Code. 30461197
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461197 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30461197 Supplier LoGiCal Supplier No. LGCFOR1564.00

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

Isoleucine (L-Isoleucine)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Isoleucine
CAS 73-32-5
Analyte or Component Names L-Isoleucine
Shipping Condition Room Temperature
Molecular Formula C6 H13 N O2
Certifications/Compliance ISO 9001
Synonym Isoleucine, L-Isoleucine (9 CI, ACI), Isoleucine, L- (8 CI), (2 S,3 S)-2-Amino-3-methylpentanoic acid, (2 S,3 S)-α-Amino-β-methyl-n-valeric acid, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-, Pentanoic acid, 2-amino-3-methyl-, (2 S,3 S)-
InChI Formula InChI=1 S/C6H13NO2/c1-3-4(2)5(7)6(8)9/h4-5 H,3,7H2,1-2H3,(H,8,9)/t4-,5-/m0/s1
SMILES CC[C@H](C)[C@H](N)C(=O)O
IUPAC Name (2 S,3 S)-2-amino-3-methylpentanoic acid
Industry Type Forensics
Molecular Weight (g/mol) 131.17
Quantity 10 mg
Formula Weight 131.0946 g/mol
Show More Show Less
Product Title
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.