missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl 2-(4-Isobutylphenyl)propanoate, Mikromol™

Product Code. 17942806
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942806 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17942806 Supplier Mikromol Supplier No. MM0002.23

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Isopropyl 2-(4-Isobutylphenyl)propanoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl 2-(4-Isobutylphenyl)propanoate
CAS 64622-17-9
Analyte or Component Names Isopropyl 2-(4-Isobutylphenyl)propanoate
Shipping Condition Room Temperature
Molecular Formula C16 H24 O2
Certifications/Compliance ISO 17025
Synonym Isopropyl 2-(4-Isobutylphenyl)propanoate
InChI Formula InChI=1S/C16H24O2/c1-11(2)10-14-6-8-15(9-7-14)13(5)16(17)18-12(3)4/h6-9,11-13H,10H2,1-5H3
SMILES CC(C)Cc1ccc(cc1)C(C)C(=O)OC(C)C
IUPAC Name propan-2-yl 2-[4-(2-methylpropyl)phenyl]propanoate
Molecular Weight (g/mol) 248.36
Quantity 100 mg
Formula Weight 248.1776 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.