missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CaM Kinase II Monoclonal specifically detects CaM Kinase II in Human∣Mouse∣Rat samples. It is validated for Flow Cytometry∣Immunocytochemistry/Immunofluorescence∣Immunohistochemistry∣Western Blot.
Specifications
Specifications
| Antigen | CaM Kinase II |
| Applications | Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | SR1956 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:100, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Formulation | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gene Alias | Calcium/calmodulin-dependent protein kinase;Calcium/calmodulin-dependent protein kinase type II subunit beta;CAM2;CAMK2;CAMK2B;CAMKB;CaMKIIbeta;MRD54 |
| Host Species | Rabbit |
| Immunogen | A synthesized peptide derived from human CaM Kinase II (Uniprot #: Q9UQM7, Q13554, Q13555, Q13557) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?