missing translation for 'onlineSavingsMsg'
Learn More

Isopropyl Salicylate, Mikromol™

Product Code. 17983646
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983646 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17983646 Supplier Mikromol Supplier No. MM0045.17
 Restricted Item
Estimated Shipment: 31-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Isopropyl Salicylate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isopropyl Salicylate
CAS 607-85-2
Analyte or Component Names Isopropyl Salicylate
Shipping Condition Room Temperature
Molecular Formula C10 H12 O3
Certifications/Compliance ISO 9001
Synonym Isopropyl Salicylate, 2-Hydroxybenzoic acid 1-methylethyl ester, Salicylic acid, isopropyl ester (6CI,7CI,8CI), 2-Hydroxybenzoic acid isopropyl ester, Isopropyl o-hydroxybenzoate, Isopropyl salicylate
InChI Formula InChI=1S/C10H12O3/c1-7(2)13-10(12)8-5-3-4-6-9(8)11/h3-7,11H,1-2H3
SMILES CC(C)OC(=O)c1ccccc1O
IUPAC Name propan-2-yl 2-hydroxybenzoate
Molecular Weight (g/mol) 180.2
Quantity 100 mg
Formula Weight 180.0786 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.