missing translation for 'onlineSavingsMsg'
Learn More

Isosorbide Mononitrate 1.0 mg/ml in Dimethyl Sulfoxide, LoGiCal

Product Code. 30464209
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30464209 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30464209 Supplier LoGiCal Supplier No. LGCAMP0502.0029
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Isosorbide Mononitrate
CAS 16051-77-7
Analyte or Component Names Isosorbide Mononitrate
Shipping Condition Room Temperature
Molecular Formula C6 H9 N O6
Certifications/Compliance ISO 17034
Synonym Isosorbide Dinitrate, Diluted Imp. C (EP), Isosorbide Mononitrate, Isosorbide Dinitrate Imp. C (EP), Isosorbide 5-Nitrate, 1,4:3,6-Dianhydro-D-glucitol 5-nitrate, D-1,4:3,6-Dianhydro-D-glucitol 5-nitrate, Furo[3,2-b]furan, D-glucitol deriv., AHR 4698, BM 22-145, Conpin, Conpin Retardkaps, Corangin, Corangin SR, Dilavenil, Duride, Elan, Elantan, Elantan Long, Elantan Retard, Geomatrix, IS 5MN, ISMN, Imdur, Imdur 60, Imdur Durules, Imodur, Imtrate, Ismexin, Ismo, Ismo 20, Ismox, Isomon, Isomonat, Isomonit, Isopen 20, Isosorbide 5-mononitrate, Isosorbide 5-nitrate, Isosorbide mononitrate, Iturol, Medocor, Momo Mack, Monicor, Monis, Monit, Monit 20, Monizid, Mono Corax, Mono Corax Retard, Mono-Cedocard, Mono-Mack, Mono-Sanorania, Monoclair, Monocord 20, Monocord 40, Monocord 50SR, Monodur Durules, Monoket, Monoket OD, Monoket Retard, Monolong, Monolong 40, Monolong 60, Mononit, Mononit 20, Mononit 40, Mononit Retard 50, Monopront, Monosorb, Monosorbitrate, Monosordil, Monotrate, Monovas, Multitab, Nitramin, Olicard, Ormox, Pentacard, Pentacard 20, Vasotrate, Isosorbide Dinitrate Impurity C
InChI Formula InChI=1S/C6H9NO6/c8-3-1-11-6-4(13-7(9)10)2-12-5(3)6/h3-6,8H,1-2H2/t3-,4+,5+,6+/m0/s1
Solution Type Single Solution
SMILES O[C@H]1CO[C@@H]2[C@@H](CO[C@H]12)O[N+](=O)[O-]
IUPAC Name [(3S,3aR,6R,6aS)-3-hydroxy-2,3,3a,5,6,6a-hexahydrofuro[3,2-b]furan-6-yl] nitrate
Solvent or Matrix Dimethyl Sulfoxide
Industry Type Clinical Testing
Molecular Weight (g/mol) 191.14
Quantity 1 mL
Formula Weight 191.043 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.