missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ITCH/AIP4 Monoclonal antibody specifically detects ITCH/AIP4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | ITCH/AIP4 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | S03-4I6 |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Gene Alias | AIF4, AIP4dJ468O1.1 (atrophin 1 interacting protein 4 (AIP4)), atrophin-1 interacting protein 4, Atrophin-1-interacting protein 4, dJ468O1.1, EC 6.3.2, EC 6.3.2.-, Itch, itchy (mouse homolog) E3 ubiquitin protein ligase, itchy E3 ubiquitin protein ligase homolog (mouse), itchy homolog E3 ubiquitin protein ligase, NAPP1E3 ubiquitin-protein ligase Itchy homolog, NFE2-associated polypeptide 1, ubiquitin protein ligase ITCH |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide of human ITCH/AIP4 (Uniprot # Q96J02) |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?