missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITK Antibody (5G6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-37578
This item is not returnable.
View return policy
Description
ITK Monoclonal specifically detects ITK in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| ITK | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.03% Sodium Azide | |
| ITK | |
| Purified recombinant fragment of human ITK expressed in E. Coli. | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 5G6 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| EC 2.7.10, EC 2.7.10.2, EMTMGC126257, homolog of mouse T-cell itk/tsk, IL2-inducible T-cell kinase, Kinase EMT, LYKMGC126258, PSCTK2, T-cell-specific kinase, tyrosine-protein kinase ITK/TSK, Tyrosine-protein kinase Lyk | |
| Mouse | |
| 72 kDa | |
| 0.1 mL | |
| Protein Kinase | |
| 3702 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction