missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ITK Antibody (5G6), Novus Biologicals™
Mouse Monoclonal Antibody
205.00 € - 552.00 €
Specifications
| Antigen | ITK |
|---|---|
| Clone | 5G6 |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686075
|
Novus Biologicals
NBP2-37578-0.025ml |
0.025 mL |
205.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18117657
|
Novus Biologicals
NBP2-37578 |
0.1 mL |
552.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ITK Monoclonal specifically detects ITK in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| ITK | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200 - 1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| EC 2.7.10, EC 2.7.10.2, EMTMGC126257, homolog of mouse T-cell itk/tsk, IL2-inducible T-cell kinase, Kinase EMT, LYKMGC126258, PSCTK2, T-cell-specific kinase, tyrosine-protein kinase ITK/TSK, Tyrosine-protein kinase Lyk | |
| ITK | |
| IgG1 | |
| Unpurified |
| 5G6 | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Protein Kinase | |
| Ascites with 0.03% Sodium Azide | |
| 3702 | |
| Purified recombinant fragment of human ITK expressed in E. Coli. | |
| Primary | |
| 72 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title