missing translation for 'onlineSavingsMsg'
Learn More

JNK/JIP3 Antibody, Novus Biologicals™

Product Code. 18020176 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18020176 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18020176 Supplier Novus Biologicals Supplier No. NB1001246

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

JNK/JIP3 Polyclonal specifically detects JNK/JIP3 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen JNK/JIP3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 4 - 6 ug/ml, Immunohistochemistry-Paraffin 4 - 6 ug/ml, Peptide ELISA Detection limit 1:4000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_055948
Gene Alias C-jun-amino-terminal kinase interacting protein 3, C-Jun-amino-terminal kinase-interacting protein 3, DKFZp762N1113, homolog of Drosophila Sunday driver 2, JIP-3, JIP3FLJ00027, JNK MAP kinase scaffold protein 3, JNK/SAPK-associated protein-1, JNK/stress-activated protein kinase-associated protein 1, JNK-interacting protein 3, JSAP1, KIAA1066SYD2, mitogen-activated protein kinase 8 interacting protein 3, Mitogen-activated protein kinase 8-interacting protein 3, syd
Gene Symbols MAPK8IP3
Host Species Goat
Immunogen Peptide with sequence MEIQMDEGGGVV-C corresponding to N-Terminus according to NP_055948.2, NP_001035529.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23162
Test Specificity This antibody is expected to recognize both reported Human isoforms of this protein (as represented by NP_055948.2 and NP_001035529.1).
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.