missing translation for 'onlineSavingsMsg'
Learn More

JNK1/2/3, p Thr183 Antibody (SR1788), Novus Biologicals™

Product Code. 18657964 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18657964 100 μg 100µL
18675764 50 μg 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18657964 Supplier Novus Biologicals Supplier No. NBP321529100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

JNK1/2/3 Monoclonal antibody specifically detects JNK1/2/3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen JNK1/2/3
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone SR1788
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8JNK, protein kinase JNK1, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1
Host Species Rabbit
Immunogen A synthesized peptide derived from human phospho-JNK1/2/3 [Thr183, Thr183, Thr221] (Uniprot #: P45983, P45984, P53779)
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Breast Cancer, Cancer, Cell Cycle and Replication, Diabetes Research, Hypoxia, MAP Kinase Signaling, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5599
Target Species Human, Mouse, Rat
Content And Storage Store at -20 degrees C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.