missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNK1, p Thr183 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NB100-82009-0.025ML
This item is not returnable.
View return policy
Description
JNK1 Polyclonal specifically detects JNK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| JNK1 (p Thr183) | |
| Polyclonal | |
| Unconjugated | |
| PBS [pH 7.4] 150mM NaCl and 50% glycerol (without Mg2+ and Ca2+) with 0.02% Sodium Azide | |
| c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8JNK, protein kinase JNK1, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1 | |
| Rabbit | |
| 50 kDa | |
| 0.025 mL | |
| Cancer, Hypoxia, MAP Kinase Signaling, p53 Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| 5599 | |
| Mouse, Rat, Human | |
| IgG |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
| 1.0 mg/mL | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| P45984/P53779 | |
| MAPK8 | |
| The antiserum was produced against synthesized phosphopeptide derived from human JNK2/3/SAPK around the phosphorylation site of threonine 183 (M-M-TP-P-Y). | |
| Affinity Purified | |
| RUO | |
| Primary | |
| SAPK/JNK (specific to Phospho-Thr183) detects endogenous levels of SAPK/JNK only when phosphorylated at threonine 183. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction