missing translation for 'onlineSavingsMsg'
Learn More
Learn More
JNK1, p Thr183 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
278.00 € - 458.00 €
Specifications
| Antigen | JNK1 (p Thr183) |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 |
| Applications | Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18223755
|
Novus Biologicals
NB100-82009-0.025ML |
0.025 mL |
278.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18769733
|
Novus Biologicals
NB100-82009 |
0.05 mL |
458.00 €
0.05mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
JNK1 Polyclonal specifically detects JNK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| JNK1 (p Thr183) | |
| Western Blot 1:500-1:1000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin 1:50-1:100 | |
| Polyclonal | |
| Rabbit | |
| Cancer, Hypoxia, MAP Kinase Signaling, p53 Pathway, Phospho Specific, Protein Kinase, Signal Transduction, Wnt Signaling Pathway | |
| P45984/P53779 | |
| 5599 | |
| IgG | |
| Affinity Purified | |
| 50 kDa |
| 1.0 mg/mL | |
| Immunohistochemistry (Paraffin), Immunohistochemistry, Western Blot | |
| Unconjugated | |
| RUO | |
| Mouse, Rat, Human | |
| c-Jun N-terminal kinase 1, EC 2.7.11, EC 2.7.11.24, JNK1 alpha protein kinase, JNK1 beta protein kinase, JNK1JNK1A2, JNK-46, JUN N-terminal kinase, MAP kinase 8, MAPK 8, mitogen-activated protein kinase 8, mitogen-activated protein kinase 8 isoform JNK1 alpha1, mitogen-activated protein kinase 8 isoform JNK1 beta2, PRKM8JNK, protein kinase JNK1, SAPK1JNK21B1/2, Stress-activated protein kinase 1, Stress-activated protein kinase JNK1 | |
| MAPK8 | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title