missing translation for 'onlineSavingsMsg'
Learn More

Kaempferol, Mikromol™

Product Code. 18072510
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18072510 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18072510 Supplier Mikromol Supplier No. MM1542.00
 Restricted Item
Estimated Shipment: 20-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Kaempferol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Kaempferol
CAS 520-18-3
Analyte or Component Names Kaempferol
Shipping Condition Room Temperature
Molecular Formula C15 H10 O6
Certifications/Compliance ISO 9001
Synonym Kaempferol
InChI Formula InChI=1S/C15H10O6/c16-8-3-1-7(2-4-8)15-14(20)13(19)12-10(18)5-9(17)6-11(12)21-15/h1-6,16-18,20H
SMILES OC1=C(Oc2cc(O)cc(O)c2C1=O)c3ccc(O)cc3
IUPAC Name 3,5,7-trihydroxy-2-(4-hydroxyphenyl)chromen-4-one
Molecular Weight (g/mol) 286.24
Quantity 250 mg
Formula Weight 286.0477 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.