missing translation for 'onlineSavingsMsg'
Learn More

KAT3B/p300 Antibody (RW128) - BSA Free, Novus Biologicals™

Product Code. 18019545 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18019545 0.025 mg 25µg
18777303 0.1 mg 0.1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18019545 Supplier Novus Biologicals Supplier No. NB100507SS

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 7 publications

KAT3B/p300 Monoclonal specifically detects KAT3B/p300 in Human, Mouse, Rat, Mustelid, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Flow (Intracellular).
TRUSTED_SUSTAINABILITY

Specifications

Antigen KAT3B/p300
Applications Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Monoclonal
Clone RW128
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:250-1:500, Flow Cytometry, Immunocytochemistry/Immunofluorescence 2-5ug/mL, Immunoprecipitation 1:10-1:500, Flow (Intracellular) 1ug/mL
Gene Alias E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2
Gene Symbols EP300
Host Species Mouse
Immunogen Fusion protein containing residues 1572-2371 of human KAT3B/p300. [UniProt# Q0947]
Molecular Weight of Antigen 300 kDa
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, HIF Target Genes, Hypoxia, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 2033
Target Species Human, Mouse, Rat, Mustelid, Primate
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.