missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPAG4L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-56960
This item is not returnable.
View return policy
Description
SPAG4L Polyclonal specifically detects SPAG4L in Human samples. It is validated for Western Blot.
Specifications
| SPAG4L | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| dJ726C3.1, MGC33594, Sad1 and UNC84 domain containing 5, SPAG4L, Sperm-associated antigen 4-like protein, SUN domain-containing protein 5, testis and spermatogenesis related gene 4, Testis and spermatogenesis-related gene 4 protein, TSARG4sperm associated antigen 4-like | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 140732 | |
| Human | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q8TC36 | |
| SUN5 | |
| Synthetic peptides corresponding to SPAG4L(sperm associated antigen 4-like) The peptide sequence was selected from the N terminal of SPAG4L. Peptide sequence MPRSSRSPGDPGALLEDVAHNPRPRRIAQRGRNTSRMAEDTSPNMNDNIL. | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction