missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Kinesin 5A Polyclonal specifically detects Kinesin 5A in Human samples. It is validated for Immunoprecipitation, Western Blot.
Specifications
Specifications
| Applications | Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | DyLight 488 |
| Dilution | Western Blot |
| Immunogen | The immunogen this antibody was made to, maps to a region between residue 982 to 1032 of human Kinesin Family Member 5A using the numbering given in entry NP_004975.2 (GeneID 3798). |
| Content And Storage | Store at 4°C in the dark. |
| Product Type | Antibody |
| Isotype | IgG |
| Product Line | Novus |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Politique de confidentialité.
Vous avez repéré une opportunité d'amélioration ?