missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
KLC2 Polyclonal antibody specifically detects KLC2 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
Specifications
| Antigen | KLC2 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Biotin |
| Formulation | PBS |
| Gene Alias | FLJ12387, kinesin light chain 2, KLC 2 |
| Host Species | Rabbit |
| Immunogen | kinesin light chain 2. (Uniprot# Q9H0B6) |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?