missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KLF12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
450.00 € - 708.00 €
Specifications
| Antigen | KLF12 |
|---|---|
| Dilution | Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18684145
|
Novus Biologicals
NBP3-21391-25ul |
25 μg |
450.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653245
|
Novus Biologicals
NBP3-21391-100ul |
100 μg |
708.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KLF12 Polyclonal antibody specifically detects KLF12 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| KLF12 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| DNA replication Transcription Translation and Splicing | |
| PBS, pH 7.2, 40% glycerol | |
| 11278 | |
| IgG | |
| Affinity purified |
| Immunohistochemistry 1:20 - 1:50, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| AP-2rep, AP-2rep transcription factor, AP2REPAP-2 repressor, HSPC122, KLF12 zinc finger transcriptional repressor, Krueppel-like factor 12, Kruppel-like factor 12, Transcriptional repressor AP-2rep | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: SSPTVITSVSSASSSSTVLTPGPLVASASGVGGQQFLHIIHPVPPSSPMNLQSNKLSHVHRIPVVVQSVPVVYTAVRSPGNV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title