missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ KMT2D Antibody (3C8E11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27155-0.025ml
This item is not returnable.
View return policy
Description
KMT2D Monoclonal antibody specifically detects KMT2D in Human samples. It is validated for Flow Cytometry,ELISA,Western Blot
Specifications
| KMT2D | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| AAD10, ALRALL1-related protein, CAGL114, EC 2.1.1.43, histone-lysine N-methyltransferase MLL2, KMT2D, Lysine N-methyltransferase 2B, MLL4, myeloid/lymphoid or mixed-lineage leukemia 2, Myeloid/lymphoid or mixed-lineage leukemia protein 2, TNRC21, trinucleotide repeat containing 21 | |
| Purified recombinant fragment of human KMT2D (AA: 445-599) expressed in E. Coli. | |
| 0.025 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, ELISA, Western Blot | |
| 3C8E11 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 8085 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction