missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ KMT2D Antibody (3C8E11), Novus Biologicals™
Mouse Monoclonal Antibody
310.00 € - 534.00 €
Specifications
| Antigen | KMT2D |
|---|---|
| Clone | 3C8E11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 |
| Applications | Flow Cytometry, ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30029107
|
Novus Biologicals™
NBP3-27155-0.025ml |
0.025 mL |
310.00 €
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30039885
|
Novus Biologicals™
NBP3-27155-0.1ml |
0.1 mL |
534.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KMT2D Monoclonal antibody specifically detects KMT2D in Human samples. It is validated for Flow Cytometry,ELISA,Western BlotSpecifications
| KMT2D | |
| 1 mg/mL | |
| Flow Cytometry, ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| AAD10, ALRALL1-related protein, CAGL114, EC 2.1.1.43, histone-lysine N-methyltransferase MLL2, KMT2D, Lysine N-methyltransferase 2B, MLL4, myeloid/lymphoid or mixed-lineage leukemia 2, Myeloid/lymphoid or mixed-lineage leukemia protein 2, TNRC21, trinucleotide repeat containing 21 | |
| Purified recombinant fragment of human KMT2D (AA: 445-599) expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 3C8E11 | |
| Western Blot 1:500 - 1:2000, Flow Cytometry 1:200 - 1:400, ELISA 1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 8085 | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title