missing translation for 'onlineSavingsMsg'
Learn More

KMT2D Antibody (CL5346) [DyLight 755], Novus Biologicals™

Product Code. 30525774 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30525774 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30525774 Supplier Novus Biologicals Supplier No. NBP343877IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

KMT2D Monoclonal antibody specifically detects KMT2D in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen KMT2D
Applications Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone CL5346
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias AAD10, ALRALL1-related protein, CAGL114, EC 2.1.1.43, histone-lysine N-methyltransferase MLL2, KMT2D, Lysine N-methyltransferase 2B, MLL4, myeloid/lymphoid or mixed-lineage leukemia 2, Myeloid/lymphoid or mixed-lineage leukemia protein 2, TNRC21, trinucleotide repeat containing 21
Host Species Mouse
Immunogen This antibody was generated using a recombinant protein sequence of O14686, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8085
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.