Learn More
Invitrogen™ KSR1 Recombinant Rabbit Monoclonal Antibody (23GB6290), Invitrogen™
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA553956
 View more versions of this product
Product Code. 30089034
  • 562.00 € / 100µL
Description
Part of a multiprotein signaling complex which promotes phosphorylation of Raf family members and activation of downstream MAP kinases. Independently of its kinase activity, acts as MAP2K1/MEK1 and MAP2K2/MEK2-dependent allosteric activator of BRAF; upon binding to MAP2K1/MEK1 or MAP2K2/MEK2, dimerizes with BRAF and promotes BRAF-mediated phosphorylation of MAP2K1/MEK1 and/or MAP2K2/MEK2 (PubMed:29433126). Promotes activation of MAPK1 and/or MAPK3, both in response to EGF and to cAMP. Its kinase activity is unsure. Some protein kinase activity has been detected in vitro, however the physiological relevance of this activity is unknown. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KSR1
Recombinant Monoclonal
0.375 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8IVT5
KSR1
A synthesized peptide derived from human KSR1 (880-923AA).
100 μL
Primary
Human, Rat
Antibody
IgG
Flow Cytometry, Western Blot, Immunocytochemistry
23GB6290
Unconjugated
KSR1
AW492498; B-KSR1; CAP kinase; D11Bhm183e; D11Bhm184e; hb; HB protein; kinase KSR1; Kinase suppressor of Ras 1; Ksr; KSR1; mKSR1; OTTHUMP00000162931; protein Hb; RSU2
Rabbit
Affinity chromatography
RUO
360573, 8844
-20°C