missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KSR1 Recombinant Rabbit Monoclonal Antibody (23GB6290), Invitrogen™
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

207.00 € - 562.00 €

Specifications

Antigen KSR1
Clone 23GB6290
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30089034
GREENER_CHOICE
View Documents
Invitrogen™
MA553956
100 μL
562.00 €
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30088095
View Documents
Invitrogen™
MA553957
20 μL
207.00 €
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

KSR1 Recombinant Monoclonal antibody specifically detects KSR1 in Human, Rat samples. It is validated for Flow Cytometry, Western Blot
Specifications

Specifications

KSR1
Flow Cytometry, Western Blot, Immunocytochemistry
Unconjugated
Rabbit
Human, Rat
Q8IVT5
360573, 8844
IgG
Affinity chromatography
Antibody
23GB6290
Recombinant Monoclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
AW492498; B-KSR1; CAP kinase; D11Bhm183e; D11Bhm184e; hb; HB protein; kinase KSR1; Kinase suppressor of Ras 1; Ksr; KSR1; mKSR1; OTTHUMP00000162931; protein Hb; RSU2
A synthesized peptide derived from human KSR1 (880-923AA).
Primary
-20°C
KSR1
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.