missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Kv4.2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30798-100ug
This item is not returnable.
View return policy
Description
Kv4.2 Polyclonal antibody specifically detects Kv4.2 in Rat samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Kv4.2 | |
| Polyclonal | |
| Western Blot : 1:1000, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| potassium voltage-gated channel, Shal-related subfamily, member 2, RK5, voltage-gated potassium channel Kv4.2, voltage-sensitive potassium channel | |
| Rabbits were immunized with a peptide representing amino acids 453-470 of rat Kv4.2 conjugated to KLH. | |
| 100 μg | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 3751 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction