missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Kv4.2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | Kv4.2 |
|---|---|
| Dilution | Western Blot : 1:1000, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation |
| Applications | Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066934
|
Novus Biologicals™
NBP3-30798-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067151
|
Novus Biologicals™
NBP3-30798-100ug |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kv4.2 Polyclonal antibody specifically detects Kv4.2 in Rat samples. It is validated for Immunoprecipitation,Western Blot,Immunocytochemistry/ ImmunofluorescenceSpecifications
| Kv4.2 | |
| Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| Phosphate buffered saline | |
| 3751 | |
| IgG | |
| Ammonium sulfate precipitation |
| Western Blot : 1:1000, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation | |
| Polyclonal | |
| Purified | |
| RUO | |
| Rat | |
| potassium voltage-gated channel, Shal-related subfamily, member 2, RK5, voltage-gated potassium channel Kv4.2, voltage-sensitive potassium channel | |
| Rabbits were immunized with a peptide representing amino acids 453-470 of rat Kv4.2 conjugated to KLH. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title