missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine, Mikromol™

Product Code. 17988954
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17988954 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17988954 Supplier Mikromol Supplier No. MM0024.03
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

L-Cystine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Cystine
CAS 56-89-3
Analyte or Component Names L-Cystine
Shipping Condition Room Temperature
Molecular Formula C6 H12 N2 O4 S2
Certifications/Compliance ISO 17034
Synonym L-Cystine, L-Cystine (9CI, ACI), Cystine, Cystine, L- (8CI), (-)-Cystine, 3,3′-Dithiobis(2-aminopropanoic acid), L-(-)-Cystine, L-Alanine, 3,3′-dithiobis-, L-Cysteine disulfide, L-Cystin, [R-(R*,R*)]-3,3′-Dithiobis[2-aminopropanoic acid], Bis(β-amino-β-carboxyethyl) disulfide, Cystine acid, Dicysteine, l-Cystine, NSC 13203, Oxidized L-cysteine, Propanoic acid, 3,3′-dithiobis[2-amino-, [R-(R*,R*)]-, β,β′-Diamino-β,β′-dicarboxydiethyl disulfide, β,β′-Dithiodialanine, Acetylcysteine Imp. A (EP), Acetylcysteine Impurity A, Tyrosine Impurity C
InChI Formula InChI=1S/C6H12N2O4S2/c7-3(5(9)10)1-13-14-2-4(8)6(11)12/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12)/t3-,4-/m0/s1
SMILES N[C@@H](CSSC[C@H](N)C(=O)O)C(=O)O
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
Molecular Weight (g/mol) 240.3
Quantity 100 mg
Formula Weight 240.0238 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.